2-[2-(Methylsulphonyl)phenyl]ethylamine hydrochloride
Catalog # APOSOR470914-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.731 g/mol
Storage Temperature:Ambient
CAS Number:2006277-03-6
Formula:C₉H₁₃NO₂S·ClH
MDL Number:MFCD27986816