2-[4-(Pyridin-3-ylmethoxy)phenyl]ethylamine
Catalog # APOSOR470908-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.29 g/mol
Storage Temperature:Ambient
CAS Number:1343316-76-6
Formula:C₁₄H₁₆N₂O
MDL Number:MFCD09044240