2-[4-(Pyrrolidin-3-yloxy)phenyl]ethylamine
Catalog # APOSOR470906-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.284 g/mol
Storage Temperature:Ambient
CAS Number:1000507-30-1
Formula:C₁₂H₁₈N₂O
MDL Number:MFCD09924818