2-[4-(Tetrahydropyran-2-yloxy)phenyl]ethylamine
Catalog # APOSOR470905-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.295 g/mol
Storage Temperature:Ambient
CAS Number:1000550-74-2
Formula:C₁₃H₁₉NO₂
MDL Number:MFCD09924455