2-(4-Cyclobutoxyphenyl)ethylamine
Catalog # APOSOR470902-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.269 g/mol
Storage Temperature:Ambient
CAS Number:1554206-32-4
Formula:C₁₂H₁₇NO
MDL Number:MFCD23107401