Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate
Catalog # APOSOR470866-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate
Catalog # APOSOR470866-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.241 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1453423-57-8
Formula:C₁₁H₁₅NO₄
MDL Number:MFCD27935386