Order Entry
Czech Republic
ContactUsLinkComponent
 
(R)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Catalog # APOSOR470786-5G
Supplier:  Apollo Scientific
CAS Number:  
(R)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Catalog # APOSOR470786-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :472.169 g/mol
Storage Temperature:Ambient
CAS Number:117745-45-6
Formula:C₂₂H₁₆Br₂O₂
MDL Number:MFCD06654379