(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Catalog # APOSOR470785-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:472.169 g/mol
Storage Temperature:Ambient
CAS Number:117745-41-2
Formula:C₂₂H₁₆Br₂O₂
MDL Number:MFCD06654379