Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Iodoethyl)-2-methyl-1,3-dioxolane
2-(2-Iodoethyl)-2-methyl-1,3-dioxolane
Catalog # APOSOR470781-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Iodoethyl)-2-methyl-1,3-dioxolane
Catalog # APOSOR470781-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.055 g/mol
Storage Temperature:Ambient
CAS Number:53750-51-9
Formula:C₆H₁₁IO₂
MDL Number:MFCD27500653