1'-Boc-spiro[indane-1,4'-piperidine]
Catalog # APOSOR470772-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:287.397 g/mol
Storage Temperature:Ambient
CAS Number:148835-99-8
Formula:C₁₈H₂₅NO₂
MDL Number:MFCD10700122