Order Entry
Czech Republic
ContactUsLinkComponent
2-(Hydroxymethyl)-2-phenyl-1,3-propanediol
2-(Hydroxymethyl)-2-phenyl-1,3-propanediol
Catalog # APOSOR470765-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(Hydroxymethyl)-2-phenyl-1,3-propanediol
Catalog # APOSOR470765-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :182.216 g/mol
Storage Temperature:Refrigerator
CAS Number:4704-99-8
Formula:C₁₀H₁₄O₃
MDL Number:MFCD18642376