2-(2-Thienyl)imidazo[1,2-a]pyridine-7-carboxylic acid
Catalog # APOSOR470752-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.269 g/mol
Storage Temperature:Ambient
CAS Number:928003-75-2
Formula:C₁₂H₈N₂O₂S
MDL Number:MFCD08164315