3-Chloro-5-(hydroxymethyl)benzoic acid
Catalog # APOSOR470740-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.592 g/mol
Storage Temperature:Ambient
CAS Number:153203-56-6
Formula:C₈H₇ClO₃
MDL Number:MFCD20658596