3-Cyano-5-(methoxycarbonyl)benzoic acid
Catalog # APOSOR470706-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.167 g/mol
Storage Temperature:Ambient
CAS Number:126739-90-0
Formula:C₁₀H₇NO₄
MDL Number:MFCD08458980