Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl (1S,2S)-2-[[(S)-1-phenylethyl]amino]cyclopentanecarboxylate hydrochloride
Ethyl (1S,2S)-2-[[(S)-1-phenylethyl]amino]cyclopentanecarboxylate hydrochloride
Catalog # APOSOR470689-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl (1S,2S)-2-[[(S)-1-phenylethyl]amino]cyclopentanecarboxylate hydrochloride
Catalog # APOSOR470689-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.82 g/mol
Storage Temperature:Ambient
CAS Number:359586-65-5
Formula:C₁₆H₂₃NO₂·ClH
MDL Number:MFCD23703876