Order Entry
Czech Republic
ContactUsLinkComponent
(R)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione
(R)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione
Catalog # APOSOR470686-5G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione
Catalog # APOSOR470686-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :255.654 g/mol
Storage Temperature:Ambient
CAS Number:338786-76-8
Formula:C₁₁H₁₀ClNO₄
MDL Number:MFCD26523289