Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3-(4-Methyl-1-piperazinyl)propanoate
Methyl 3-(4-Methyl-1-piperazinyl)propanoate
Catalog # APOSOR470661-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-(4-Methyl-1-piperazinyl)propanoate
Catalog # APOSOR470661-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :186.251 g/mol
Storage Temperature:Ambient
CAS Number:33544-40-0
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD08062551