Methyl 4-bromo-2-(4-methyl-1-piperazinyl)benzoate
Catalog # APOSOR470640-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:313.19 g/mol
Storage Temperature:Ambient
CAS Number:1434128-45-6
Formula:C₁₃H₁₇BrN₂O₂
MDL Number:MFCD24335260