Order Entry
Czech Republic
ContactUsLinkComponent
 
Ethyl 4-(4-Methyl-1-piperazinyl)butanoate
Catalog # APOSOR470587-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-(4-Methyl-1-piperazinyl)butanoate
Catalog # APOSOR470587-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :214.305 g/mol
Storage Temperature:Ambient
CAS Number:487008-51-5
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD01876880