Order Entry
Czech Republic
ContactUsLinkComponent
4-(3-Pyridyl)-3-butyn-1-ol
4-(3-Pyridyl)-3-butyn-1-ol
Catalog # APOSOR470563-5G
Supplier:  Apollo Scientific
CAS Number:  
4-(3-Pyridyl)-3-butyn-1-ol
Catalog # APOSOR470563-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :147.174 g/mol
Storage Temperature:Ambient
CAS Number:138487-20-4
Formula:C₉H₉NO
MDL Number:MFCD12808175