Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl 3-Bromobenzylphosphonate
Diethyl 3-Bromobenzylphosphonate
Diethyl 3-Bromobenzylphosphonate
Diethyl 3-Bromobenzylphosphonate
APOSOR470556-5G
:
Apollo Scientific
:
Diethyl 3-Bromobenzylphosphonate
APOSOR470556-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
307.121 g/mol
:
Ambient
:
128833-03-4
:
C₁₁H₁₆BrO₃P
:
MFCD07772009