4-(5-Nitrobenzimidazol-2-yl)aniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1385694-71-2
:
Formula:C₁₃H₁₀N₄O₂·ClH
MDL Number:MFCD22423151
:290.705 g/mol
Storage Temperature:Ambient
Frequently Bought Together