Order Entry
Czech Republic
ContactUsLinkComponent
 
N-Cbz-4-(1,3-dioxolan-2-yl)aniline
Catalog # APOSOR470545-5G
Supplier:  Apollo Scientific
CAS Number:  
N-Cbz-4-(1,3-dioxolan-2-yl)aniline
Catalog # APOSOR470545-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :299.321 g/mol
Storage Temperature:Ambient
CAS Number:1224886-44-5
Formula:C₁₇H₁₇NO₄
MDL Number:MFCD22375700