Methyl 1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470537-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:291.299 g/mol
Storage Temperature:Ambient
CAS Number:1385694-51-8
Formula:C₁₅H₁₇NO₅
MDL Number:MFCD22423144