Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470528-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470528-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :262.301 g/mol
Storage Temperature:Ambient
CAS Number:1385694-68-7
Formula:C₁₅H₁₈O₄
MDL Number:MFCD22423129