(S)-1-Boc-2-propylpiperazine hydrochloride
Catalog # APOSOR470454-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.792 g/mol
Storage Temperature:Ambient
CAS Number:1217478-55-1
Formula:C₁₂H₂₄N₂O₂·ClH
MDL Number:MFCD11101260