Order Entry
Czech Republic
ContactUsLinkComponent
(R)-1-Boc-2-propylpiperazine hydrochloride
(R)-1-Boc-2-propylpiperazine hydrochloride
Catalog # APOSOR470453-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-1-Boc-2-propylpiperazine hydrochloride
Catalog # APOSOR470453-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.792 g/mol
Storage Temperature:Ambient
CAS Number:1217449-00-7
Formula:C₁₂H₂₄N₂O₂·ClH
MDL Number:MFCD11101218