Ethyl 2-(2-Benzimidazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Catalog # APOSOR470445-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:299.285 g/mol
Storage Temperature:Ambient
CAS Number:195251-35-5
Formula:C₁₄H₁₃N₅O₃
MDL Number:MFCD22205839