5-(4-Methoxyphenyl)imidazolidine-2,4-dione
Catalog # APOSOR470423-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:206.198 g/mol
Storage Temperature:Ambient
CAS Number:6617-78-3
Formula:C₁₀H₁₀N₂O₃
MDL Number:MFCD00219347