1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Catalog # APOSOR470409-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:248.274 g/mol
Storage Temperature:Ambient
CAS Number:887978-56-5
Formula:C₁₄H₁₆O₄
MDL Number:MFCD06410573