Order Entry
Czech Republic
ContactUsLinkComponent
 
4-(Aminomethyl)-4-(3,4-dichlorophenyl)cyclohexanol
Catalog # APOSOR470367-5G
Supplier:  Apollo Scientific
CAS Number:  
4-(Aminomethyl)-4-(3,4-dichlorophenyl)cyclohexanol
Catalog # APOSOR470367-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :274.186 g/mol
Storage Temperature:Ambient
CAS Number:887978-44-1
Formula:C₁₃H₁₇Cl₂NO
MDL Number:MFCD06410570