[8-(3-Methylphenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887978-87-2
:
Formula:C₁₆H₂₃NO₂
MDL Number:MFCD06410583
:261.359 g/mol
Storage Temperature:Ambient
Frequently Bought Together