3-(Boc-amino)-3-(3-chlorophenyl)-1-propanol
Catalog # APOSOR470354-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:285.766 g/mol
Storage Temperature:Ambient
CAS Number:1314406-54-6
Formula:C₁₄H₂₀ClNO₃
MDL Number:MFCD18907505