2-[(Cyclopropylcarbonyl)amino]benzoic acid
Catalog # APOSOR470307-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:205.21 g/mol
Storage Temperature:Ambient
CAS Number:40057-21-4
Formula:C₁₁H₁₁NO₃
MDL Number:MFCD02602218