Methyl 2,6-Dihydroxypyridine-4-carboxylate hydrochloride
Catalog # APOSOR470299-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:205.596 g/mol
Storage Temperature:Ambient
CAS Number:1314406-42-2
Formula:C₇H₇NO₄·ClH
MDL Number:MFCD19442784