Zadání objednávky
Czech Republic
ContactUsLinkComponent
2-Methyl-1,3-dioxolane-2-acetamide
2-Methyl-1,3-dioxolane-2-acetamide
Catalog # APOSOR470298-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-1,3-dioxolane-2-acetamide
Catalog # APOSOR470298-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :145.156 g/mol
Storage Temperature:Ambient
CAS Number:70829-14-0
Formula:C₆H₁₁NO₃
MDL Number:MFCD19442782