Order Entry
Czech Republic
ContactUsLinkComponent
3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
Catalog # APOSOR470286-5G
Supplier:  Apollo Scientific
CAS Number:  
3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
Catalog # APOSOR470286-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :314.357 g/mol
Storage Temperature:Ambient
CAS Number:1265892-01-0
Formula:C₁₃H₁₈N₂O₅S
MDL Number:MFCD18433028