5-(2-Methoxyphenyl)thiophene-2-carbaldehyde
Catalog # APOSOR470275-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:218.272 g/mol
Storage Temperature:Ambient
CAS Number:479243-27-1
Formula:C₁₂H₁₀O₂S
MDL Number:MFCD06410189