Order Entry
Czech Republic
ContactUsLinkComponent
 
7-[4-(Methylsulphonyl)phenyl]-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470241-5G
Supplier:  Apollo Scientific
CAS Number:  
7-[4-(Methylsulphonyl)phenyl]-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSOR470241-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :280.343 g/mol
Storage Temperature:Ambient
CAS Number:1272755-84-6
Formula:C₁₃H₁₆N₂O₃S
MDL Number:MFCD18651753