6-Nitrospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
Catalog # APOSOR470220-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:247.25 g/mol
Storage Temperature:Ambient
CAS Number:158439-85-1
Formula:C₁₂H₁₃N₃O₃
MDL Number:MFCD18651722