Methyl 4-(2-Ethoxy-2-oxoethylamino)butanoate
Catalog # APOSOR470157-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:203.236 g/mol
Storage Temperature:Ambient
CAS Number:1183267-12-0
Formula:C₉H₁₇NO₄
MDL Number:MFCD12148722