Order Entry
Czech Republic
ContactUsLinkComponent
4,5-Isoxazoledicarbonitrile
4,5-Isoxazoledicarbonitrile
Catalog # APOSOR470135-5G
Supplier:  Apollo Scientific
CAS Number:  
4,5-Isoxazoledicarbonitrile
Catalog # APOSOR470135-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :119.081 g/mol
Storage Temperature:Ambient
CAS Number:1256633-32-5
Formula:C₅HN₃O
MDL Number:MFCD18070995