Order Entry
Czech Republic
ContactUsLinkComponent
(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
Catalog # APOSOR470117-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-N6-[2-(trans-4-Aminocyclohexyl)ethyl]-N6-propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
Catalog # APOSOR470117-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :336.538 g/mol
Storage Temperature:Ambient
CAS Number:1393886-88-8
Formula:C₁₈H₃₂N₄S
MDL Number:MFCD18911728