(5S,18S)-1,22-Diamino-7,16-dioxo-6,8,15,17-tetraazadocosane-5,18-dicarboxylic acid
Catalog # APOSOR470114-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:460.568 g/mol
Storage Temperature:Ambient
CAS Number:1417789-80-0
Formula:C₂₀H₄₀N₆O₆
MDL Number:MFCD16883073