2-Chloro-4-(4-methyl-1-piperazinyl)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1092770-25-6
:
Formula:C₁₂H₁₅ClN₂O₂
MDL Number:MFCD13191626
:254.713 g/mol
Storage Temperature:Ambient
Frequently Bought Together