Methyl 3-(1-Imidazolyl)benzoate
Catalog # APOSOR470028-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:202.209 g/mol
Storage Temperature:Ambient
CAS Number:335255-85-1
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD12198856