3-(4-Methyl-1-piperazinylsulphonyl)benzoic acid
Catalog # APOSOR470025-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:284.331 g/mol
Storage Temperature:Ambient
CAS Number:380339-63-9
Formula:C₁₂H₁₆N₂O₄S
MDL Number:MFCD02725037