Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile
4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile
Catalog # APOSOR46747-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile
Catalog # APOSOR46747-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :177.59 g/mol
Storage Temperature:Ambient
CAS Number:1263420-00-3
Formula:C₈H₄ClN₃
MDL Number:MFCD28053422