Amino(4-methoxyphenyl)acetonitrile hydrochloride
Catalog # APOSOR46701-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.649 g/mol
Storage Temperature:Ambient
CAS Number:5537-50-8
Formula:C₉H₁₀N₂O·ClH
MDL Number:MFCD27959559