
2-[(Dimethylamino)methylene]indan-1-one
Catalog # APOSOR46645-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.238 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:150542-55-5
Formula:C₁₂H₁₃NO
MDL Number:MFCD00219908